Product Details

SNP ID
rs4454083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:161687516 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACCAGTAGGTGTTCTGGGGACTC[C/T]GTTCCAGGTACAGATGGCTGTAGCA
Phenotype
MIM: 137143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRA6 PubMed Links

Gene Details

Gene
GABRA6
Gene Name
gamma-aminobutyric acid type A receptor alpha6 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000811.2 Intron NP_000802.2

View Full Product Details