Product Details

SNP ID
rs3751122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64185649 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGCGAGAGGGAAAGCAACCCAGA[G/T]GCCCCGCAGCCGCCGGCGACACAGC
Phenotype
MIM: 605063
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STIP1 PubMed Links

Gene Details

Gene
STIP1
Gene Name
stress induced phosphoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282652.1 378 UTR 5 NP_001269581.1
NM_001282653.1 378 Intron NP_001269582.1
NM_006819.2 378 Intron NP_006810.1

View Full Product Details