Product Details

SNP ID
rs35571359
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:114606130 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AAAAAACTTGGACACACTTCTGTAA[C/T]ACATCTTGAATATAGTCTCATCTGT
Phenotype
MIM: 601291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UGT8 PubMed Links

Gene Details

Gene
UGT8
Gene Name
UDP glycosyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128174.2 Intron NP_001121646.1
NM_001322112.1 Intron NP_001309041.1
NM_001322113.1 Intron NP_001309042.1
NM_001322114.1 Intron NP_001309043.1
NM_003360.4 Intron NP_003351.2

View Full Product Details