Product Details
- SNP ID
-
rs8107999
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:21403434 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAGACAAATAGAGAAACATATTTT[C/T]ATATTCATTGAGCAGCTCATTGTTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF493
PubMed Links
Gene Details
- Gene
- ZNF493
- Gene Name
- zinc finger protein 493
View Full Product Details