Product Details

SNP ID
rs7854533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:134865008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCACCAGCTGTGTGAGGAGGCAG[C/T]GGGCGTACAGGGCGCTGGCGGGCAG
Phenotype
MIM: 601624
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCN2 PubMed Links

Gene Details

Gene
FCN2
Gene Name
ficolin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004108.2 Intron NP_004099.2
NM_015837.2 Intron NP_056652.1
XM_006717015.3 Intron XP_006717078.1
XM_011518392.2 Intron XP_011516694.1
Gene
LOC101448202
Gene Name
uncharacterized LOC101448202
There are no transcripts associated with this gene.

View Full Product Details