Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001320793.1 | 4997 | Intron | NP_001307722.1 | ||
NM_004379.4 | 4997 | UTR 3 | NP_004370.1 | ||
NM_134442.4 | 4997 | UTR 3 | NP_604391.1 | ||
XM_011510645.1 | 4997 | UTR 3 | XP_011508947.1 | ||
XM_011510646.2 | 4997 | UTR 3 | XP_011508948.1 | ||
XM_011510647.2 | 4997 | UTR 3 | XP_011508949.1 | ||
XM_011510648.2 | 4997 | Intron | XP_011508950.1 | ||
XM_011510649.2 | 4997 | UTR 3 | XP_011508951.1 | ||
XM_011510650.2 | 4997 | UTR 3 | XP_011508952.1 | ||
XM_011510651.2 | 4997 | UTR 3 | XP_011508953.1 | ||
XM_017003399.1 | 4997 | Intron | XP_016858888.1 | ||
XM_017003400.1 | 4997 | UTR 3 | XP_016858889.1 | ||
XM_017003401.1 | 4997 | UTR 3 | XP_016858890.1 |