Product Details

SNP ID
rs4486677
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8516520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTGCCAGCCTCATTAATTAATTA[G/T]CAGGGAAATGCAAATGCAACTCACA
Phenotype
MIM: 609964
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CLEC4D PubMed Links

Gene Details

Gene
CLEC4D
Gene Name
C-type lectin domain family 4 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080387.4 Intron NP_525126.2
XM_011520632.2 Intron XP_011518934.1
XM_011520633.2 Intron XP_011518935.1

View Full Product Details