Product Details

SNP ID
rs7155799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90869968 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGACTGCCTTTTTACCCATTTCAT[C/T]GCCTGTCGAATCCTCCTCAAGAAGA
Phenotype
MIM: 603607
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPS6KA5 PubMed Links

Gene Details

Gene
RPS6KA5
Gene Name
ribosomal protein S6 kinase A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322227.1 4622 UTR 3 NP_001309156.1
NM_001322228.1 4622 UTR 3 NP_001309157.1
NM_001322229.1 4622 UTR 3 NP_001309158.1
NM_001322230.1 4622 UTR 3 NP_001309159.1
NM_001322231.1 4622 UTR 3 NP_001309160.1
NM_001322232.1 4622 UTR 3 NP_001309161.1
NM_001322233.1 4622 UTR 3 NP_001309162.1
NM_001322234.1 4622 UTR 3 NP_001309163.1
NM_001322235.1 4622 UTR 3 NP_001309164.1
NM_001322236.1 4622 UTR 3 NP_001309165.1
NM_001322237.1 4622 UTR 3 NP_001309166.1
NM_001322238.1 4622 UTR 3 NP_001309167.1
NM_004755.3 4622 UTR 3 NP_004746.2
NM_182398.2 4622 Intron NP_872198.1
XM_017021786.1 4622 Intron XP_016877275.1

View Full Product Details