Product Details

SNP ID
rs10814376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36343167 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATTTTTTCCTGAATATTTTCAA[C/T]CTTTGGTTGGTTGAATACAGAGGGC
Phenotype
MIM: 612488
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF38 PubMed Links

Gene Details

Gene
RNF38
Gene Name
ring finger protein 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022781.4 Intron NP_073618.3
NM_194328.2 Intron NP_919309.1
NM_194329.2 Intron NP_919310.1
NM_194330.2 Intron NP_919311.1
NM_194332.2 Intron NP_919313.1
XM_005251366.3 Intron XP_005251423.1
XM_005251367.3 Intron XP_005251424.1
XM_005251368.3 Intron XP_005251425.1
XM_006716721.3 Intron XP_006716784.1
XM_011517712.2 Intron XP_011516014.1
XM_011517713.2 Intron XP_011516015.1
XM_017014294.1 Intron XP_016869783.1
XM_017014295.1 Intron XP_016869784.1
XM_017014296.1 Intron XP_016869785.1
XM_017014297.1 Intron XP_016869786.1

View Full Product Details