Product Details
- SNP ID
-
rs5753659
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31497627 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCCCCTAAGTGTTCAGTGAAAGGA[A/G]GAGTTCCATGTCTCTCACTTTATAT
- Phenotype
-
MIM: 607445
MIM: 612765
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EIF4ENIF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111281097] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EIF4ENIF1
- Gene Name
- eukaryotic translation initiation factor 4E nuclear import factor 1
There are no transcripts associated with this gene.
- Gene
- SFI1
- Gene Name
- SFI1 centrin binding protein
View Full Product Details