Product Details

SNP ID
rs2339803
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:179662627 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATGGATGTCTTTGTGTCATTTGCCA[C/T]GTGTACCCTAAAATCCTGCTTTTCT
Phenotype
MIM: 603591
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP13 PubMed Links

Gene Details

Gene
USP13
Gene Name
ubiquitin specific peptidase 13 (isopeptidase T-3)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003940.2 Intron NP_003931.2
XM_011513269.1 Intron XP_011511571.1
XM_017007425.1 Intron XP_016862914.1
XM_017007426.1 Intron XP_016862915.1
XM_017007427.1 Intron XP_016862916.1

View Full Product Details