Product Details

SNP ID
rs11967559
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:142076496 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGAAGCAGGGCTTGCTGTTTAAA[A/G]CTACCACTCCACAGGAAAACAAAAT
Phenotype
MIM: 162341
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NMBR PubMed Links
Additional Information
For this assay, SNP(s) [rs146678853] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NMBR
Gene Name
neuromedin B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324307.1 Intron NP_001311236.1
NM_001324308.1 Intron NP_001311237.1
NM_002511.3 Intron NP_002502.2
XM_011535846.1 Intron XP_011534148.1
XM_017010901.1 Intron XP_016866390.1
XM_017010902.1 Intron XP_016866391.1

View Full Product Details