Product Details
- SNP ID
-
rs12934479
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75000691 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGGCTGCTGTACGGAGGGCCCAG[C/T]CGGCGAGTCTCGCGGGGGCTTTGCA
- Phenotype
-
MIM: 612060
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNRF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs553566194] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNRF1
- Gene Name
- zinc and ring finger 1, E3 ubiquitin protein ligase
View Full Product Details