Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001195131.1 | 2978 | Intron | NP_001182060.1 | ||
XM_017010141.1 | 2978 | Intron | XP_016865630.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001080481.1 | 2978 | UTR 3 | NP_001073950.1 | ||
XM_005267169.3 | 2978 | UTR 3 | XP_005267226.1 | ||
XM_005267170.4 | 2978 | UTR 3 | XP_005267227.1 | ||
XM_005267172.3 | 2978 | UTR 3 | XP_005267229.1 | ||
XM_005267175.3 | 2978 | Intron | XP_005267232.1 | ||
XM_011536200.2 | 2978 | UTR 3 | XP_011534502.1 | ||
XM_017011381.1 | 2978 | UTR 3 | XP_016866870.1 | ||
XM_017011382.1 | 2978 | UTR 3 | XP_016866871.1 | ||
XM_017011383.1 | 2978 | UTR 3 | XP_016866872.1 | ||
XM_017011384.1 | 2978 | UTR 3 | XP_016866873.1 | ||
XM_017011385.1 | 2978 | Intron | XP_016866874.1 | ||
XM_017011386.1 | 2978 | Intron | XP_016866875.1 |