Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001178137.1 | 3299 | UTR 3 | NP_001171608.1 | ||
NM_014912.4 | 3299 | UTR 3 | NP_055727.3 | ||
XM_005269630.4 | 3299 | UTR 3 | XP_005269687.1 | ||
XM_006717714.3 | 3299 | UTR 3 | XP_006717777.1 | ||
XM_006717715.3 | 3299 | UTR 3 | XP_006717778.1 | ||
XM_011539514.2 | 3299 | UTR 3 | XP_011537816.1 | ||
XM_011539515.2 | 3299 | UTR 3 | XP_011537817.1 | ||
XM_011539517.2 | 3299 | UTR 3 | XP_011537819.1 | ||
XM_011539518.2 | 3299 | Intron | XP_011537820.1 | ||
XM_011539519.2 | 3299 | UTR 3 | XP_011537821.1 | ||
XM_011539520.2 | 3299 | UTR 3 | XP_011537822.1 | ||
XM_011539522.2 | 3299 | Intron | XP_011537824.1 | ||
XM_011539523.2 | 3299 | Intron | XP_011537825.1 | ||
XM_017015927.1 | 3299 | UTR 3 | XP_016871416.1 |