Product Details

SNP ID
rs2230004
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:128463608 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGAGTAATTCCAGAAGAAACTGCCA[C/T]AGCTGGATTGGTCCACTGCCTAGAA
Phenotype
MIM: 164720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETS1 PubMed Links

Gene Details

Gene
ETS1
Gene Name
ETS proto-oncogene 1, transcription factor
There are no transcripts associated with this gene.

View Full Product Details