Product Details
- SNP ID
-
rs11202914
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:88970454 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAGGCCCCTGCTTGTTGAGGAGGG[C/T]GTTCAGTTAGTGCTGAGACTACCTG
- Phenotype
-
MIM: 102620
MIM: 134637
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACTA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113631043] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACTA2
- Gene Name
- actin, alpha 2, smooth muscle, aorta
- Gene
- FAS
- Gene Name
- Fas cell surface death receptor
View Full Product Details