Product Details

SNP ID
rs12192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120396106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCTAGTATGTGGCAGGGTGGG[A/G]GCATCTGGGTTATTTCAGCCCCTGC
Phenotype
MIM: 605547
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FSTL1 PubMed Links

Gene Details

Gene
FSTL1
Gene Name
follistatin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007085.4 1948 UTR 3 NP_009016.1
Gene
LOC100130701
Gene Name
uncharacterized LOC100130701
There are no transcripts associated with this gene.

Gene
MIR198
Gene Name
microRNA 198
There are no transcripts associated with this gene.

View Full Product Details