Product Details

SNP ID
rs17119173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56923609 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGCTATTGCTGCAGATCGCTGAA[A/C]CTGCAAAGACCACCTCAGGTTTCTG
Phenotype
MIM: 609769
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SDR9C7 PubMed Links

Gene Details

Gene
SDR9C7
Gene Name
short chain dehydrogenase/reductase family 9C, member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_148897.2 1310 UTR 3 NP_683695.1

View Full Product Details