Product Details
- SNP ID
-
rs12448948
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:25700666 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GCATAATGGATGATCTCTCTGGAAT[A/G]GATTGAACCACCGCCCCGTTCAAGT
- Phenotype
-
MIM: 604059
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HS3ST4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115717481] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HS3ST4
- Gene Name
- heparan sulfate-glucosamine 3-sulfotransferase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006040.2 |
|
Intron |
|
|
NP_006031.2 |
View Full Product Details