Product Details

SNP ID
rs7858453
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:102997565 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCAGGCTGCAGTCAGTTCCTAGAAA[A/T]CTTTCTACCATGTCTTTTATGGTCT
Phenotype
MIM: 604035
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYLC2 PubMed Links

Gene Details

Gene
CYLC2
Gene Name
cylicin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001340.4 Intron NP_001331.1

View Full Product Details