Product Details

SNP ID
rs12686532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125748349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCCGGGCAGATGGGTCCGCCT[C/T]GTTCCGGCTGCAGCTTTCGCCGCCG
Phenotype
MIM: 176312
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC51145 PubMed Links

Gene Details

Gene
LOC51145
Gene Name
uncharacterized LOC51145
There are no transcripts associated with this gene.

Gene
PBX3
Gene Name
PBX homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134778.1 151 UTR 5 NP_001128250.1
NM_006195.5 151 Intron NP_006186.1
XM_006717130.1 151 Intron XP_006717193.1
XM_006717131.1 151 Intron XP_006717194.1
XM_006717132.1 151 Intron XP_006717195.1
XM_006717133.1 151 Intron XP_006717196.1
XM_011518755.1 151 UTR 5 XP_011517057.1
XM_017014786.1 151 UTR 5 XP_016870275.1

View Full Product Details