Product Details

SNP ID
rs4318547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119182196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATCTATGGAAAATAAAAAGAA[A/G]TACAAATGGGTATCTTGTGTAAGCC
Phenotype
MIM: 602380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UPK1B PubMed Links

Gene Details

Gene
UPK1B
Gene Name
uroplakin 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006952.3 Intron NP_008883.2

View Full Product Details