Product Details
- SNP ID
-
rs13390401
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:188293419 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTACGGAGAATGTTCAAGTCATGA[C/G]TGATTTCTAGGACTAGCAGGAGACA
- Phenotype
-
MIM: 608165
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GULP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114828436] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GULP1
- Gene Name
- GULP, engulfment adaptor PTB domain containing 1
- Gene
- LINC01090
- Gene Name
- long intergenic non-protein coding RNA 1090
There are no transcripts associated with this gene.
- Gene
- MIR561
- Gene Name
- microRNA 561
There are no transcripts associated with this gene.
View Full Product Details