Product Details

SNP ID
rs10022662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:163338461 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGACTCTGACCCAGAGGCTCCCC[A/C]GTTTGTTGCTGAGTGACATCACCCA
Phenotype
MIM: 162641 MIM: 602001
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NPY1R PubMed Links

Gene Details

Gene
NPY1R
Gene Name
neuropeptide Y receptor Y1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000909.5 1494 Intron NP_000900.1
XM_005263031.3 1494 Intron XP_005263088.1
XM_011532010.2 1494 Intron XP_011530312.1
Gene
NPY5R
Gene Name
neuropeptide Y receptor Y5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317091.1 1494 Intron NP_001304020.1
NM_001317092.1 1494 Intron NP_001304021.1
NM_006174.3 1494 Intron NP_006165.1
XM_005263038.3 1494 UTR 5 XP_005263095.1
XM_011532015.2 1494 UTR 5 XP_011530317.1
XM_011532017.2 1494 UTR 5 XP_011530319.1
XM_017008255.1 1494 Intron XP_016863744.1
XM_017008256.1 1494 UTR 5 XP_016863745.1

View Full Product Details