Product Details

SNP ID
rs6628886
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:34636660 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACTGTCAGTTACACAAAAAGAAATA[C/T]GCTGTAGTCAACTGTACTGGAAATG
Phenotype
MIM: 300698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM47 PubMed Links

Gene Details

Gene
TMEM47
Gene Name
transmembrane protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031442.3 Intron NP_113630.1

View Full Product Details