Product Details

SNP ID
rs4478858
Assay Type
Functionally tested
NCBI dbSNP Submissions
69
Location
Chr.1:31411078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCATCCAGTAAGCATTCACTGC[C/T]TGACTGTGTGCCAGGCCCTGTACTG
Phenotype
MIM: 614549
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERINC2 PubMed Links

Gene Details

Gene
SERINC2
Gene Name
serine incorporator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199037.1 Intron NP_001185966.1
NM_001199038.1 Intron NP_001185967.1
NM_001199039.1 Intron NP_001185968.1
NM_018565.3 Intron NP_061035.2
NM_178865.4 Intron NP_849196.2

View Full Product Details