Product Details
- SNP ID
-
rs34151874
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:35755323 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTTTTCACTTTTTCCTGCTTGTC[A/G]CCCTCTTCCTAACTGCCCCCTGACT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ26245
PubMed Links
Gene Details
- Gene
- FLJ26245
- Gene Name
- uncharacterized LOC400533
There are no transcripts associated with this gene.
View Full Product Details