Product Details
- SNP ID
-
rs11992605
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:66955543 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCATAAAATGTTTTTAAATGCCTGG[C/T]AGATAATCCACAGTGCTCAAATAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TCF24
PubMed Links
Gene Details
- Gene
- TCF24
- Gene Name
- transcription factor 24
View Full Product Details