Product Details

SNP ID
rs35939148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81705977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTAAACTGGATTGCTTTTGAGGA[C/G]GGAGAAGATGTTATTAATAGCCTTC
Phenotype
MIM: 604375 MIM: 602375 MIM: 606794
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HGS PubMed Links

Gene Details

Gene
HGS
Gene Name
hepatocyte growth factor-regulated tyrosine kinase substrate
There are no transcripts associated with this gene.

Gene
MRPL12
Gene Name
mitochondrial ribosomal protein L12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002949.3 Intron NP_002940.2
Gene
SLC25A10
Gene Name
solute carrier family 25 member 10
There are no transcripts associated with this gene.

View Full Product Details