Product Details

SNP ID
rs11594585
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:100051409 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGCGAGTGTTGTATCAGAAAGGCC[C/G]CATTCCAGATGCAGCTACTATGAGG
Phenotype
MIM: 603103
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPN1 PubMed Links

Gene Details

Gene
CPN1
Gene Name
carboxypeptidase N subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308.2 Intron NP_001299.1

View Full Product Details