Product Details

SNP ID
rs868217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:83464303 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACAAAGTGACTATCTTACTATTA[C/T]GAAAGGATACTATTGAAGATTTTTT
Phenotype
MIM: 611143 MIM: 606769 MIM: 611983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM175A PubMed Links

Gene Details

Gene
FAM175A
Gene Name
family with sequence similarity 175 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139076.2 Intron NP_620775.2
XM_005263280.4 Intron XP_005263337.1
XM_017008694.1 Intron XP_016864183.1
Gene
HELQ
Gene Name
helicase, POLQ-like
There are no transcripts associated with this gene.

Gene
MRPS18C
Gene Name
mitochondrial ribosomal protein S18C
There are no transcripts associated with this gene.

View Full Product Details