Product Details

SNP ID
rs8056687
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:56434649 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGGTATACATCTTTAGTTTTACAA[C/T]AAACCCAAAAGTTCAAGGAACTTTG
Phenotype
MIM: 603243 MIM: 604978
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMFR PubMed Links

Gene Details

Gene
AMFR
Gene Name
autocrine motility factor receptor
There are no transcripts associated with this gene.

Gene
NUDT21
Gene Name
nudix hydrolase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007006.2 Intron NP_008937.1

View Full Product Details