Product Details

SNP ID
rs4077095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176555192 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAACAACTTTCTGCAAAAAGAGCT[T/G]CTTATTCTATGTTAGTTGGATGAGT
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
CDHR2 PubMed Links

Gene Details

Gene
CDHR2
Gene Name
cadherin related family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171976.1 Intron NP_001165447.1
NM_017675.4 Intron NP_060145.3

View Full Product Details