Product Details

SNP ID
rs17784294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102719628 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAAAACTTTATCGTAAATCACCT[A/C]GATGCTGCTGACTCAGTTTTTTCCT
Phenotype
MIM: 604695 MIM: 615570
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARL3 PubMed Links

Gene Details

Gene
ARL3
Gene Name
ADP ribosylation factor like GTPase 3
There are no transcripts associated with this gene.

Gene
SFXN2
Gene Name
sideroflexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178858.4 Intron NP_849189.1
XM_011539261.2 Intron XP_011537563.1
XM_011539262.2 Intron XP_011537564.1
XM_011539263.2 Intron XP_011537565.1
XM_011539264.2 Intron XP_011537566.1
XM_017015663.1 Intron XP_016871152.1

View Full Product Details