Product Details

SNP ID
rs2569007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:145866186 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAGAAAATATAGGGCTGATAT[A/T]TTTTTCCACCTCTAGAACCAGCTGT
Phenotype
MIM: 615762
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GRXCR2 PubMed Links

Gene Details

Gene
GRXCR2
Gene Name
glutaredoxin and cysteine rich domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080516.1 Intron NP_001073985.1
XM_017009708.1 Intron XP_016865197.1

View Full Product Details