Product Details

SNP ID
rs6503190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9252737 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCTGATGATCCAATCTTTTGAGA[A/G]ACGCTTCTCTTGGGGCCCCAAGGTT
Phenotype
MIM: 601614 MIM: 604203
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTN1 PubMed Links

Gene Details

Gene
NTN1
Gene Name
netrin 1
There are no transcripts associated with this gene.

Gene
STX8
Gene Name
syntaxin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004853.2 Intron NP_004844.1
XM_011524079.2 Intron XP_011522381.1

View Full Product Details