Product Details

SNP ID
rs11098065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:110052487 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGTGAATCAAGTGAGGAGCAATT[A/G]AAAATAACAGTTTTCATTTTACAGA
Phenotype
MIM: 611546
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELOVL6 PubMed Links

Gene Details

Gene
ELOVL6
Gene Name
ELOVL fatty acid elongase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130721.1 Intron NP_001124193.1
NM_024090.2 Intron NP_076995.1
XM_011532233.2 Intron XP_011530535.1
XM_011532234.2 Intron XP_011530536.1
XM_011532235.2 Intron XP_011530537.1

View Full Product Details