Product Details

SNP ID
rs4251926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43648343 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCACCCGCCACCACGTCCGGATTT[A/G]GCCGATTTTTAATCAAGAGGAACTA
Phenotype
MIM: 609744 MIM: 173391
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CADM4 PubMed Links

Gene Details

Gene
CADM4
Gene Name
cell adhesion molecule 4
There are no transcripts associated with this gene.

Gene
PLAUR
Gene Name
plasminogen activator, urokinase receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005376.2 Intron NP_001005376.1
NM_001005377.2 Intron NP_001005377.1
NM_001301037.1 Intron NP_001287966.1
NM_002659.3 Intron NP_002650.1
XM_005258990.4 Intron XP_005259047.1
XM_011527027.2 Intron XP_011525329.1
XM_011527028.2 Intron XP_011525330.1
XM_011527029.1 Intron XP_011525331.1
XM_011527030.2 Intron XP_011525332.1
XM_011527031.2 Intron XP_011525333.1
XM_017026872.1 Intron XP_016882361.1
XM_017026873.1 Intron XP_016882362.1

View Full Product Details