Product Details
- SNP ID
-
rs6276
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:113410675 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACGGTTCGCAAGGGTGAGGCTGGC[C/T]GGCCTGGGCAGGGAGGTGGGAAGCA
- Phenotype
-
MIM: 126450
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DRD2
PubMed Links
Gene Details
- Gene
- DRD2
- Gene Name
- dopamine receptor D2
View Full Product Details