Product Details
- SNP ID
-
rs9276427
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:32744080 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCTGTCGATATCCTCCTCACAGAA[C/T]TCCCCAAAGTACATTGTATTTGCTC
- Phenotype
-
MIM: 613503
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HLA-DQA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117735661] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HLA-DQA2
- Gene Name
- major histocompatibility complex, class II, DQ alpha 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020056.4 |
|
Intron |
|
|
NP_064440.1 |
- Gene
- MIR3135B
- Gene Name
- microRNA 3135b
There are no transcripts associated with this gene.
View Full Product Details