Product Details

SNP ID
rs1536
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:74057826 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GATGAGTTTTTTTCATCTAAGAAAT[A/G]AGAAACTCTGAGCAGCAAATAGTTG
Phenotype
MIM: 603178
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH6A1 PubMed Links

Gene Details

Gene
ALDH6A1
Gene Name
aldehyde dehydrogenase 6 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278593.1 2058 UTR 3 NP_001265522.1
NM_001278594.1 2058 UTR 3 NP_001265523.1
NM_005589.3 2058 UTR 3 NP_005580.1
XM_017021331.1 2058 UTR 3 XP_016876820.1
Gene
BBOF1
Gene Name
basal body orientation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025057.2 2058 Intron NP_079333.2
XM_005268092.3 2058 UTR 3 XP_005268149.1
XM_005268097.3 2058 UTR 3 XP_005268154.1
XM_005268098.4 2058 UTR 3 XP_005268155.1
XM_011537170.2 2058 UTR 3 XP_011535472.1
XM_011537171.2 2058 Intron XP_011535473.1
XM_011537172.1 2058 Intron XP_011535474.1
XM_011537174.2 2058 Intron XP_011535476.1
XM_011537175.2 2058 UTR 3 XP_011535477.1
XM_011537176.2 2058 UTR 3 XP_011535478.1
XM_011537177.2 2058 UTR 3 XP_011535479.1
XM_011537178.2 2058 UTR 3 XP_011535480.1
XM_011537179.2 2058 UTR 3 XP_011535481.1
XM_017021659.1 2058 Intron XP_016877148.1
XM_017021660.1 2058 Intron XP_016877149.1
XM_017021661.1 2058 Intron XP_016877150.1
XM_017021662.1 2058 Intron XP_016877151.1
XM_017021663.1 2058 Intron XP_016877152.1

View Full Product Details