Product Details
- SNP ID
-
hCV328440
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:22119056 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACAGCCCTTTCCTCCCTCCCTCT[A/C]GCAGGTGTGTGTGTGTGTGTTTTCC
- Phenotype
-
MIM: 603490
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WNT4
PubMed Links
Gene Details
- Gene
- WNT4
- Gene Name
- Wnt family member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030761.4 |
2154 |
UTR 3 |
|
|
NP_110388.2 |
View Full Product Details