Product Details
- SNP ID
-
rs9882021
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:154026161 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGTATGATTACATTTTAAGTGGAC[C/T]GTAAGAGTTAAAGTGCTCTACTTAC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF26-AS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144478367] are located under a probe and SNP(s) [rs79896515] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGEF26-AS1
- Gene Name
- ARHGEF26 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details