Product Details

SNP ID
rs5749341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31947982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTACTGGGCTAGCATTGTGTTT[C/T]AACACTGATACACCAGTTGATTACA
Phenotype
MIM: 113508
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf24 PubMed Links

Gene Details

Gene
C22orf24
Gene Name
chromosome 22 open reading frame 24
There are no transcripts associated with this gene.

Gene
YWHAH
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003405.3 Intron NP_003396.1

View Full Product Details