Product Details

SNP ID
rs5945244
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:153525306 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTAAGGCGCATGGAGTGGCCGGTTC[A/G]AAACTGAACCTACATTCCCACAGAT
Phenotype
MIM: 300014
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP2B3 PubMed Links

Gene Details

Gene
ATP2B3
Gene Name
ATPase plasma membrane Ca2+ transporting 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001344.2 Intron NP_001001344.1
NM_021949.3 Intron NP_068768.2
XM_005274689.3 Intron XP_005274746.1
XM_005274690.3 Intron XP_005274747.1
XM_005274691.3 Intron XP_005274748.1
XM_005274692.3 Intron XP_005274749.1
XM_011531174.2 Intron XP_011529476.1
XM_011531177.2 Intron XP_011529479.1
XM_017029553.1 Intron XP_016885042.1

View Full Product Details