Product Details

SNP ID
rs12559450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:123191991 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGTTTCTGGATATTACTCTCAGA[A/G]TGGCTGTTCACTCTACCCAAACTAA
Phenotype
MIM: 305915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIA3 PubMed Links

Gene Details

Gene
GRIA3
Gene Name
glutamate ionotropic receptor AMPA type subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000828.4 Intron NP_000819.3
NM_001256743.1 Intron NP_001243672.1
NM_007325.4 Intron NP_015564.4

View Full Product Details