Product Details

SNP ID
rs11965542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28272166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCTGTCAACAGTGGCTACAGCCC[A/G]AGACCCATACCAAGGAGCAGATCCT
Phenotype
MIM: 616474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGBD1 PubMed Links

Gene Details

Gene
PGBD1
Gene Name
piggyBac transposable element derived 1
There are no transcripts associated with this gene.

Gene
ZSCAN26
Gene Name
zinc finger and SCAN domain containing 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023560.3 368 Missense Mutation AAG,GAG K83E NP_001018854.2
NM_001111039.2 368 Missense Mutation AAG,GAG K83E NP_001104509.1
NM_001287421.1 368 Intron NP_001274350.1
NM_001287422.1 368 Intron NP_001274351.1
NM_152736.5 368 Intron NP_689949.3
XM_011514862.2 368 Missense Mutation AAG,GAG K83E XP_011513164.1
XM_011514864.1 368 Missense Mutation AAG,GAG K54E XP_011513166.1
XM_011514866.1 368 Intron XP_011513168.1
XM_011514867.1 368 UTR 5 XP_011513169.1
XM_011514869.1 368 UTR 5 XP_011513171.1
XM_017011264.1 368 Missense Mutation AAG,GAG K83E XP_016866753.1
XM_017011265.1 368 UTR 5 XP_016866754.1

View Full Product Details