Product Details

SNP ID
rs2238151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111774029 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGCATAGGGAGTGGTGAAACTGGC[C/T]CAGGGAAAGTCCATTGCAGCAAGAT
Phenotype
MIM: 100650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALDH2 PubMed Links

Gene Details

Gene
ALDH2
Gene Name
aldehyde dehydrogenase 2 family (mitochondrial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000690.3 Intron NP_000681.2
NM_001204889.1 Intron NP_001191818.1

View Full Product Details