Product Details
- SNP ID
-
rs3213519
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:104040753 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTTATCTTTCTAGGTTATCTTTAT[C/G]ATGATGTTATCAATGTACTGATGTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
H2BFM
PubMed Links
Gene Details
- Gene
- H2BFM
- Gene Name
- H2B histone family member M
View Full Product Details